Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB271324.1 - 1 Chr. 16 - 15705292 - 15705826 [-] on Build GRCh38 89
GenBankmRNA AK130790.1 - 1 1146 Chr. 16 - 15704410 - 15705984 [+] on Build GRCh38 89


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 484 MIR484 MIRN484, hsa-mir-484, mir-484 -61.0kb Sense Chr. - 15643294 - 15643372 on Build GRCh38
Downstream FGFR1OP N-terminal like FOPNL C16orf63, FOR20, PHSECRG2 159.7kb Anti-sense Chr. - 15865719 - 15888649 on Build GRCh38

View Full Product Details