Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- C5orf56
- Entrez Gene ID
-
441108
- Gene Name
- chromosome 5 open reading frame 56
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 132410773 - 132476044 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK025221.1 |
- |
1 - 1 |
1251 |
102 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA358892.1 |
- |
1 |
|
|
Chr. 5 - 132456445 - 132456765 [+] on Build GRCh38 |
102 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 22 member 5 |
SLC22A5 |
CDSP, OCTN2 |
-59.6kb
|
Sense |
Chr. - 132369704 - 132395614 on Build GRCh38 |
Downstream |
interferon regulatory factor 1 |
IRF1 |
IRF-1, MAR |
23.5kb
|
Anti-sense |
Chr. - 132481609 - 132490789 on Build GRCh38 |
View Full Product Details