Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK026967.1 - 1 1052 Chr. 20 - 36252560 - 36254718 [+] on Build GRCh38 70


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream AAR2 splicing factor homolog AAR2 C20orf4, CGI-23 Sense Chr. - 36236136 - 36256941 on Build GRCh38
Downstream DLG associated protein 4 DLGAP4 DAP-4, DAP4, DLP4, SAPAP-4, SAPAP4 51.6kb Sense Chr. - 36306323 - 36528637 on Build GRCh38

View Full Product Details