Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK055637.1 - 1 1248 Chr. 1 - 147175602 - 147177740 [+] on Build GRCh38 90


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream protein kinase AMP-activated non-catalytic subunit beta 2 PRKAB2 -3.0kb Anti-sense Chr. - 147155106 - 147172589 on Build GRCh38
Downstream protein disulfide isomerase family A member 3 pseudogene 1 PDIA3P1 ERp60, GRP58P, PDIA3P 111.0 Sense Chr. - 147177851 - 147179949 on Build GRCh38

View Full Product Details