Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK128509.1 - 1 2649 Chr. 7 - 16779669 - 16783274 [+] on Build GRCh38 100


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream basic leucine zipper and W2 domains 2 BZW2 HSPC028, MST017, MSTP017 -73.1kb Sense Chr. - 16646134 - 16706523 on Build GRCh38
Downstream anterior gradient 2, protein disulphide isomerase family member AGR2 AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2 8.5kb Anti-sense Chr. - 16791811 - 16805114 on Build GRCh38

View Full Product Details