The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_131187.1 | 2 - 2 | 1840 | 104 | |
GenBank mRNA | BC037918.1 | - | 3 - 3 | 1651 | 104 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BX091068.1 | - | 1 | Chr. 10 - 3750294 - 3750764 [-] on Build GRCh38 | 104 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105376501 | LOC105376501 | -192.9kb | Anti-sense | Chr. - 3532040 - 3556794 on Build GRCh38 | |
Downstream | Kruppel like factor 6 | KLF6 | BCD1, CBA1, COPEB, CPBP, GBF, PAC1, ST12, ZF9 | 25.2kb | Anti-sense | Chr. - 3775996 - 3785281 on Build GRCh38 |