Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HAUS4
- Entrez Gene ID
-
54930
- Gene Name
- HAUS augmin like complex subunit 4
- Gene Aliases
-
C14orf94
- Chromosome Location
-
Chr.14: 22946228 - 22957142 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK097980.1 |
- |
1 - 1 |
1851 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101926933 |
LOC101926933 |
|
|
Anti-sense |
Chr. - 22929609 - 22954692 on Build GRCh38 |
Downstream |
microRNA 4707 |
MIR4707 |
mir-4707 |
993.0
|
Sense |
Chr. - 22956950 - 22957029 on Build GRCh38 |
View Full Product Details