Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU621316.1 |
- |
1 |
|
|
Chr. 1 - 108690749 - 108691478 [+] on Build GRCh38 |
77 |
GenBankmRNA |
BC053353.1 |
- |
1 |
1152 |
|
Chr. 1 - 108690749 - 108692524 [-] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
HEN1 methyltransferase homolog 1 |
HENMT1 |
C1orf59, HEN1 |
-29.2kb
|
Sense |
Chr. - 108648288 - 108661526 on Build GRCh38 |
Downstream |
fibronectin type III domain containing 7 |
FNDC7 |
|
19.9kb
|
Anti-sense |
Chr. - 108712934 - 108742749 on Build GRCh38 |
View Full Product Details