Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TUBB
Entrez Gene ID
203068
Gene Name
tubulin beta class I
Gene Aliases
CDCBM6, CSCSC1, M40, OK/SW-cl.56, TUBB1, TUBB5
Chromosome Location
Chr.6: 30720201 - 30725426 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293212.1 NP_001280141.1 4 - 4 2411 82
NM_001293213.1 NP_001280142.1 5 - 5 1721 82
NM_001293214.1 NP_001280143.1 3 - 3 2195 82
NM_001293215.1 NP_001280144.1 4 - 4 2314 82
NM_001293216.1 NP_001280145.1 4 - 4 2213 82
NM_178014.3 NP_821133.1 4 - 4 2327 82
NR_120608.1 4 - 4 1728 82
GenBank mRNA AB062393.1 - 4 - 4 2155 82
AK098772.1 - 4 - 4 2172 82
BC001002.2 - 1 - 1 956 82
BC007605.1 - 4 - 4 2157 82
BC013374.2 - 4 - 4 2148 82
BC019924.1 - 4 - 4 2147 82
BC021909.1 - 4 - 4 2147 82
BC062532.1 - 4 - 4 1490 82
BQ019614.1 - 1 - 1 82

View Full Product Details