The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037.4 | NP_001028.1 | 1 - 2 | 175 | 73 |
NM_199037.4 | NP_950238.1 | 1 - 2 | 175 | 73 | |
GenBank mRNA | AK295477.1 | - | 1 - 2 | 130 | 73 |
AK313279.1 | - | 1 - 2 | 60 | 73 | |
AY391842.1 | - | 1 - 2 | 42 | 73 | |
BC067122.1 | - | 1 - 2 | 175 | 73 | |
BM805468.1 | - | 1 - 2 | 197 | 73 | |
BT019923.1 | - | 1 - 2 | 42 | 73 | |
L10338.1 | - | 1 - 2 | 139 | 73 | |
L16242.1 | - | 1 - 2 | 66 | 73 |