Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HRH4
Entrez Gene ID
59340
Gene Name
histamine receptor H4
Gene Aliases
AXOR35, BG26, GPCR105, GPRv53, H4, H4R, HH4R
Chromosome Location
Chr.18: 24460568 - 24487427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160166.1 NP_001153638.1 1 - 2 290 104

View Full Product Details