Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPYL2
Entrez Gene ID
64061
Gene Name
TSPY like 2
Gene Aliases
CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX
Chromosome Location
Chr.X: 53082327 - 53088546 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022117.3 NP_071400.1 6 - 7 2061 90
XM_017029727.1 XP_016885216.1 5 - 6 2226 90
GenBank mRNA AB015345.1 - 6 - 7 1301 90
AF254794.1 - 6 - 7 1800 90
AF273046.1 - 6 - 7 2051 90
AK292462.1 - 6 - 7 2054 90
AL713652.1 - 6 - 7 2040 90
AY040871.1 - 6 - 7 2050 90
BC001566.1 - 3 - 4 858 90
BC024270.1 - 6 - 7 2041 90

View Full Product Details