The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000330.3 | NP_000321.1 | 5 - 6 | 554 | 135 |
GenBank mRNA | AF014459.1 | - | 5 - 6 | 534 | 135 |
AL049684.1 | - | 4 - 5 | 1010 | 135 |