Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR10AD1
Entrez Gene ID
121275
Gene Name
olfactory receptor family 10 subfamily AD member 1
Gene Aliases
OR10AD1P, OR12-1
Chromosome Location
Chr.12: 48202339 - 48203292 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004134.1 NP_001004134.1 1 - 1 250 76
GenBank mRNA BC136757.1 - 1 - 1 345 76
BC136758.1 - 1 - 1 345 76

View Full Product Details