The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303255.1 | NP_001290184.1 | 3 - 4 | 237 | 99 |
NM_199340.3 | NP_955372.2 | 5 - 6 | 3341 | 99 | |
XM_011524776.1 | XP_011523078.1 | 2 - 3 | 780 | 99 | |
XM_011524777.1 | XP_011523079.1 | 2 - 3 | 780 | 99 | |
XM_011524778.2 | XP_011523080.2 | 3 - 4 | 305 | 99 | |
XM_017024593.1 | XP_016880082.1 | 3 - 4 | 3170 | 99 | |
XM_017024594.1 | XP_016880083.1 | 3 - 4 | 3170 | 99 | |
XM_017024595.1 | XP_016880084.1 | 3 - 4 | 3170 | 99 | |
XM_017024596.1 | XP_016880085.1 | 3 - 4 | 3170 | 99 | |
XM_017024597.1 | XP_016880086.1 | 3 - 4 | 3170 | 99 | |
XM_017024599.1 | XP_016880088.1 | 4 - 5 | 2542 | 99 | |
XM_017024600.1 | XP_016880089.1 | 3 - 4 | 3170 | 99 | |
XM_017024601.1 | XP_016880090.1 | 2 - 3 | 482 | 99 | |
XM_017024603.1 | XP_016880092.1 | 2 - 3 | 780 | 99 | |
XM_017024604.1 | XP_016880093.1 | 2 - 3 | 780 | 99 | |
XM_017024605.1 | XP_016880094.1 | 2 - 3 | 780 | 99 | |
XM_017024607.1 | XP_016880096.1 | 2 - 3 | 85 | 99 | |
XM_017024608.1 | XP_016880097.1 | 2 - 3 | 1102 | 99 | |
GenBank mRNA | AB011135.2 | - | 4 - 5 | 3206 | 99 |
AK294371.1 | - | 3 - 4 | 225 | 99 | |
AK315945.1 | - | 2 - 3 | 813 | 99 | |
BC043145.1 | - | 2 - 3 | 2683 | 99 | |
BI560889.1 | - | 2 - 3 | 122 | 99 | |
JQ790599.1 | - | 2 - 3 | 235 | 99 | |
JQ790603.1 | - | 2 - 3 | 235 | 99 | |
JQ790607.1 | - | 2 - 3 | 235 | 99 | |
JQ790608.1 | - | 2 - 3 | 235 | 99 | |
JQ790612.1 | - | 2 - 3 | 235 | 99 | |
JQ790613.1 | - | 2 - 3 | 235 | 99 | |
JQ790632.1 | - | 2 - 3 | 235 | 99 | |
JQ790680.1 | - | 2 - 3 | 223 | 99 | |
JQ790704.1 | - | 2 - 3 | 235 | 99 | |
JQ790705.1 | - | 2 - 3 | 235 | 99 | |
JQ790706.1 | - | 2 - 3 | 235 | 99 | |
JQ790728.1 | - | 2 - 3 | 237 | 99 | |
JQ790731.1 | - | 2 - 3 | 237 | 99 | |
JQ790733.1 | - | 2 - 3 | 237 | 99 | |
JQ790734.1 | - | 2 - 3 | 245 | 99 | |
JQ790737.1 | - | 2 - 3 | 237 | 99 | |
JQ790747.1 | - | 2 - 3 | 237 | 99 | |
JQ790751.1 | - | 2 - 3 | 237 | 99 | |
JQ790753.1 | - | 2 - 3 | 237 | 99 | |
JQ790759.1 | - | 2 - 3 | 237 | 99 | |
JQ790760.1 | - | 2 - 3 | 235 | 99 | |
JQ790761.1 | - | 2 - 3 | 237 | 99 | |
JQ790762.1 | - | 2 - 3 | 238 | 99 | |
JQ790765.1 | - | 2 - 3 | 237 | 99 | |
JQ790766.1 | - | 2 - 3 | 237 | 99 | |
JQ790767.1 | - | 2 - 3 | 237 | 99 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | JQ790562.1 | - | 2 - 3 | 235 | 99 |
JQ790566.1 | - | 2 - 3 | 235 | 99 | |
JQ790577.1 | - | 2 - 3 | 235 | 99 | |
JQ790578.1 | - | 2 - 3 | 235 | 99 | |
JQ790579.1 | - | 2 - 3 | 235 | 99 | |
JQ790580.1 | - | 2 - 3 | 235 | 99 | |
JQ790581.1 | - | 2 - 3 | 235 | 99 | |
JQ790688.1 | - | 2 - 3 | 235 | 99 | |
JQ790690.1 | - | 2 - 3 | 235 | 99 | |
JQ790691.1 | - | 2 - 3 | 235 | 99 | |
JQ790692.1 | - | 2 - 3 | 235 | 99 | |
JQ790693.1 | - | 2 - 3 | 235 | 99 | |
JQ790694.1 | - | 2 - 3 | 235 | 99 | |
JQ790726.1 | - | 2 - 3 | 237 | 99 | |
JQ790727.1 | - | 2 - 3 | 237 | 99 | |
JQ790729.1 | - | 2 - 3 | 237 | 99 | |
JQ790730.1 | - | 2 - 3 | 237 | 99 | |
JQ790738.1 | - | 2 - 3 | 237 | 99 | |
JQ790743.1 | - | 2 - 3 | 237 | 99 | |
JQ790744.1 | - | 2 - 3 | 236 | 99 | |
JQ790755.1 | - | 2 - 3 | 237 | 99 | |
JQ790757.1 | - | 2 - 3 | 237 | 99 | |
JQ790758.1 | - | 2 - 3 | 237 | 99 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006607.2 | NP_001006608.2 | 2 - 3 | 2678 | 99 |
XM_011524841.2 | XP_011523143.1 | 2 - 3 | 2731 | 99 | |
XM_011524842.2 | XP_011523144.1 | 2 - 3 | 2731 | 99 | |
XM_011524843.2 | XP_011523145.1 | 2 - 3 | 2731 | 99 | |
XM_011524844.2 | XP_011523146.1 | 2 - 3 | 2731 | 99 | |
XM_011524848.2 | XP_011523150.1 | 2 - 3 | 2731 | 99 | |
XM_011524849.1 | XP_011523151.1 | 2 - 3 | 2731 | 99 | |
XM_011524850.2 | XP_011523152.1 | 2 - 3 | 2731 | 99 | |
GenBank mRNA | AK126994.1 | - | 2 - 3 | 1360 | 99 |
BC144427.1 | - | 2 - 3 | 341 | 99 | |
BC144428.1 | - | 2 - 3 | 341 | 99 | |
JQ790563.1 | - | 2 - 3 | 235 | 99 | |
JQ790564.1 | - | 2 - 3 | 235 | 99 | |
JQ790568.1 | - | 2 - 3 | 235 | 99 | |
JQ790571.1 | - | 2 - 3 | 235 | 99 | |
JQ790576.1 | - | 2 - 3 | 235 | 99 | |
JQ790584.1 | - | 2 - 3 | 235 | 99 | |
JQ790606.1 | - | 2 - 3 | 235 | 99 | |
JQ790610.1 | - | 2 - 3 | 235 | 99 | |
JQ790617.1 | - | 2 - 3 | 235 | 99 | |
JQ790626.1 | - | 2 - 3 | 235 | 99 | |
JQ790628.1 | - | 2 - 3 | 235 | 99 | |
JQ790665.1 | - | 2 - 3 | 235 | 99 | |
JQ790700.1 | - | 2 - 3 | 235 | 99 | |
JQ790703.1 | - | 2 - 3 | 211 | 99 | |
JQ790735.1 | - | 2 - 3 | 237 | 99 | |
JQ790739.1 | - | 2 - 3 | 237 | 99 | |
JQ790740.1 | - | 2 - 3 | 237 | 99 | |
JQ790750.1 | - | 2 - 3 | 237 | 99 | |
JQ790764.1 | - | 2 - 3 | 237 | 99 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | JQ790698.1 | - | 2 - 3 | 235 | 99 |