The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007532.2 | NP_001007533.1 | 1 - 1 | 350 | 88 |
GenBank mRNA | AY179170.1 | - | 1 - 1 | 320 | 88 |
BC130319.1 | - | 1 - 1 | 350 | 88 | |
BC130321.1 | - | 1 - 1 | 350 | 88 |