The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009608.2 | NP_001009608.1 | 6 - 7 | 582 | 89 |
XM_017027647.1 | XP_016883136.1 | 5 - 6 | 4084 | 89 | |
XM_017027648.1 | XP_016883137.1 | 7 - 8 | 1535 | 89 | |
XM_017027649.1 | XP_016883138.1 | 7 - 8 | 728 | 89 | |
XM_017027650.1 | XP_016883139.1 | 7 - 8 | 1495 | 89 | |
XM_017027651.1 | XP_016883140.1 | 4 - 5 | 533 | 89 | |
XM_017027652.1 | XP_016883141.1 | 6 - 7 | 1484 | 89 | |
GenBank mRNA | BC020787.1 | - | 6 - 7 | 582 | 89 |
BC026094.1 | - | 6 - 7 | 586 | 89 |