The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035267.1 | NP_001030344.1 | 1 - 2 | 210 | 72 |
GenBank mRNA | BC017820.1 | - | 1 - 2 | 165 | 72 |
BC066328.1 | - | 1 - 2 | 122 | 72 | |
BC082759.1 | - | 1 - 2 | 150 | 72 |