Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRKAG2
Entrez Gene ID
51422
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 2
Gene Aliases
AAKG, AAKG2, CMH6, H91620p, WPWS
Chromosome Location
Chr.7: 151556114 - 151877231 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040633.1 NP_001035723.1 12 - 13 1742 102
NM_001304527.1 NP_001291456.1 10 - 11 1346 102
NM_001304531.1 NP_001291460.1 9 - 10 764 102
NM_016203.3 NP_057287.2 12 - 13 2011 102
NM_024429.1 NP_077747.1 8 - 9 836 102
XM_005250002.3 XP_005250059.1 12 - 13 1877 102
XM_005250004.3 XP_005250061.1 12 - 13 1823 102
XM_005250006.4 XP_005250063.1 10 - 11 1576 102
XM_006716021.2 XP_006716084.1 12 - 13 1414 102
XM_011516282.1 XP_011514584.1 12 - 13 1411 102
XM_011516283.1 XP_011514585.1 12 - 13 1414 102
XM_011516284.1 XP_011514586.1 12 - 13 1411 102
XM_011516285.1 XP_011514587.1 9 - 10 1256 102
XM_011516286.1 XP_011514588.1 8 - 9 777 102
XM_011516287.1 XP_011514589.1 11 - 12 1487 102
XM_017012268.1 XP_016867757.1 12 - 13 1820 102
XM_017012269.1 XP_016867758.1 12 - 13 1625 102
XM_017012270.1 XP_016867759.1 12 - 13 1401 102
XM_017012271.1 XP_016867760.1 12 - 13 1820 102
XM_017012272.1 XP_016867761.1 12 - 13 1398 102
XM_017012273.1 XP_016867762.1 10 - 11 1561 102
XM_017012274.1 XP_016867763.1 9 - 10 1253 102
XM_017012275.1 XP_016867764.1 11 - 12 1750 102
XM_017012276.1 XP_016867765.1 12 - 13 1846 102
XM_017012277.1 XP_016867766.1 8 - 9 777 102
XM_017012278.1 XP_016867767.1 11 - 12 1487 102
XM_017012279.1 XP_016867768.1 11 - 12 1753 102
XM_017012280.1 XP_016867769.1 11 - 12 1750 102
XM_017012281.1 XP_016867770.1 11 - 12 1776 102
XM_017012282.1 XP_016867771.1 10 - 11 1196 102
GenBank mRNA AB025580.1 - 8 - 9 749 102
AF087875.1 - 8 - 9 797 102
AJ249976.1 - 12 - 13 1490 102
AK001887.1 - 8 - 9 836 102
AK301123.1 - 10 - 11 1295 102
AK314646.1 - 8 - 9 798 102
BC020540.1 - 8 - 9 717 102
BC068598.1 - 12 - 13 1711 102
BT007127.1 - 8 - 9 677 102

View Full Product Details