Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKAG2
Entrez Gene ID
51422
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 2
Gene Aliases
AAKG, AAKG2, CMH6, H91620p, WPWS
Chromosome Location
Chr.7: 151556114 - 151877231 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040633.1 NP_001035723.1 6 - 7 1208 85
NM_001304527.1 NP_001291456.1 4 - 5 812 85
NM_001304531.1 NP_001291460.1 3 - 4 230 85
NM_016203.3 NP_057287.2 6 - 7 1477 85
NM_024429.1 NP_077747.1 2 - 3 302 85
XM_005250002.3 XP_005250059.1 6 - 7 1343 85
XM_005250004.3 XP_005250061.1 6 - 7 1289 85
XM_005250006.4 XP_005250063.1 4 - 5 1042 85
XM_006716021.2 XP_006716084.1 6 - 7 880 85
XM_011516282.1 XP_011514584.1 6 - 7 877 85
XM_011516283.1 XP_011514585.1 6 - 7 880 85
XM_011516284.1 XP_011514586.1 6 - 7 877 85
XM_011516285.1 XP_011514587.1 3 - 4 722 85
XM_011516286.1 XP_011514588.1 2 - 3 243 85
XM_011516287.1 XP_011514589.1 5 - 6 953 85
XM_017012268.1 XP_016867757.1 6 - 7 1286 85
XM_017012269.1 XP_016867758.1 6 - 7 1091 85
XM_017012270.1 XP_016867759.1 6 - 7 867 85
XM_017012271.1 XP_016867760.1 6 - 7 1286 85
XM_017012272.1 XP_016867761.1 6 - 7 864 85
XM_017012273.1 XP_016867762.1 4 - 5 1027 85
XM_017012274.1 XP_016867763.1 3 - 4 719 85
XM_017012275.1 XP_016867764.1 5 - 6 1216 85
XM_017012276.1 XP_016867765.1 6 - 7 1312 85
XM_017012277.1 XP_016867766.1 2 - 3 243 85
XM_017012278.1 XP_016867767.1 5 - 6 953 85
XM_017012279.1 XP_016867768.1 5 - 6 1219 85
XM_017012280.1 XP_016867769.1 5 - 6 1216 85
XM_017012281.1 XP_016867770.1 5 - 6 1242 85
XM_017012282.1 XP_016867771.1 4 - 5 662 85
GenBank mRNA AB025580.1 - 2 - 3 215 85
AF087875.1 - 2 - 3 263 85
AJ249976.1 - 6 - 7 956 85
AK001887.1 - 2 - 3 302 85
AK301123.1 - 4 - 5 761 85
AK307798.1 - 5 - 6 1024 85
AK308116.1 - 6 - 7 908 85
AK314646.1 - 2 - 3 264 85
AW664063.1 - 3 - 4 202 85
BC020540.1 - 2 - 3 183 85
BT007127.1 - 2 - 3 143 85

View Full Product Details