The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080474.1 | NP_001073943.1 | 1 - 2 | 220 | 83 |
XM_005264302.3 | XP_005264359.1 | 1 - 2 | 165 | 83 | |
GenBank mRNA | AK125975.1 | - | 1 - 2 | 220 | 83 |