The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098479.1 | NP_001091949.1 | 3 - 4 | 737 | 60 |
NM_018950.2 | NP_061823.2 | 3 - 4 | 737 | 60 | |
XM_011514564.1 | XP_011512866.1 | 3 - 4 | 643 | 60 | |
XM_017010810.1 | XP_016866299.1 | 3 - 4 | 643 | 60 | |
XM_017010811.1 | XP_016866300.1 | 3 - 4 | 643 | 60 | |
XM_017010812.1 | XP_016866301.1 | 3 - 4 | 643 | 60 | |
XM_017010813.1 | XP_016866302.1 | 3 - 4 | 643 | 60 | |
XM_017010814.1 | XP_016866303.1 | 3 - 4 | 607 | 60 | |
XM_017010815.1 | XP_016866304.1 | 2 - 3 | 532 | 60 | |
GenBank mRNA | AK096962.1 | - | 3 - 4 | 643 | 60 |
AK125274.1 | - | 3 - 4 | 389 | 60 | |
AK225185.1 | - | 3 - 4 | 641 | 60 | |
AK298704.1 | - | 3 - 4 | 643 | 60 | |
AK300090.1 | - | 3 - 4 | 676 | 60 | |
AK301488.1 | - | 3 - 4 | 624 | 60 | |
AK302420.1 | - | 4 - 5 | 607 | 60 | |
AK303670.1 | - | 2 - 3 | 699 | 60 | |
AK309156.1 | - | 3 - 4 | 643 | 60 | |
AY216682.1 | - | 3 - 4 | 601 | 60 | |
AY221102.1 | - | 3 - 4 | 601 | 60 | |
AY253269.1 | - | 3 - 4 | 613 | 60 | |
AY253270.1 | - | 3 - 4 | 613 | 60 | |
BC009260.2 | - | 3 - 4 | 669 | 60 | |
BC062991.1 | - | 3 - 4 | 617 | 60 | |
CB528356.1 | - | 4 - 5 | 559 | 60 | |
DQ367723.1 | - | 3 - 4 | 613 | 60 | |
GQ891552.1 | - | 3 - 4 | 613 | 60 |