The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102420.2 | NP_001095890.1 | 5 - 6 | 1062 | 78 |
NM_001102421.2 | NP_001095891.1 | 4 - 5 | 925 | 78 | |
NM_001278243.1 | NP_001265172.1 | 5 - 6 | 641 | 78 | |
NM_001278244.1 | NP_001265173.1 | 4 - 5 | 504 | 78 | |
NM_001278245.1 | NP_001265174.1 | 5 - 6 | 638 | 78 | |
NM_006007.3 | NP_005998.1 | 4 - 5 | 922 | 78 | |
GenBank mRNA | AF062346.1 | - | 4 - 5 | 645 | 78 |
AF062347.1 | - | 4 - 5 | 500 | 78 | |
AK025782.1 | - | 5 - 6 | 525 | 78 | |
AK290849.1 | - | 4 - 5 | 645 | 78 | |
AK307376.1 | - | 2 - 3 | 346 | 78 | |
BC011018.1 | - | 4 - 5 | 761 | 78 | |
BC027707.2 | - | 5 - 6 | 1012 | 78 | |
BC073131.1 | - | 4 - 5 | 426 | 78 | |
DA290666.1 | - | 4 - 5 | 504 | 78 |