Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ING1
Entrez Gene ID
3621
Gene Name
inhibitor of growth family member 1
Gene Aliases
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a
Chromosome Location
Chr.13: 110712623 - 110721074 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198218.2 NP_937861.1 1 - 2 327 135
GenBank mRNA AF149722.1 - 1 - 2 130 135
BG703874.1 - 1 - 2 332 135

View Full Product Details