The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003897.3 | NP_003888.2 | 1 - 2 | 241 | 96 |
GenBank mRNA | AF083421.1 | - | 1 - 2 | 215 | 96 |
AK222588.1 | - | 1 - 2 | 238 | 96 | |
AK314162.1 | - | 1 - 2 | 242 | 96 | |
BC000844.2 | - | 1 - 2 | 225 | 96 | |
BC005080.1 | - | 1 - 2 | 227 | 96 | |
BM915952.1 | - | 1 - 2 | 246 | 96 | |
BT006703.1 | - | 1 - 2 | 209 | 96 | |
DA893782.1 | - | 1 - 2 | 242 | 96 | |
S81914.1 | - | 1 - 2 | 227 | 96 | |
Y14551.1 | - | 1 - 2 | 220 | 96 |