The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142348.1 | NP_001135820.1 | 1 - 2 | 306 | 75 |
GenBank mRNA | DA109496.1 | - | 1 - 2 | 287 | 75 |