The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271186.1 | NP_001258115.1 | 4 - 5 | 1123 | 90 |
NM_001271187.1 | NP_001258116.1 | 4 - 5 | 458 | 90 | |
NM_021183.4 | NP_067006.3 | 2 - 3 | 1051 | 90 | |
XM_006724775.2 | XP_006724838.1 | 3 - 4 | 781 | 90 | |
XM_011531376.1 | XP_011529678.1 | 4 - 5 | 1191 | 90 | |
GenBank mRNA | AK124801.1 | - | 1 - 2 | 361 | 90 |
AL049685.2 | - | 1 - 2 | 283 | 90 | |
AY298955.1 | - | 2 - 3 | 1051 | 90 | |
BC003403.1 | - | 1 - 2 | 337 | 90 | |
BC051467.1 | - | 2 - 3 | 1038 | 90 | |
BI767215.1 | - | 3 - 4 | 346 | 90 | |
BX647223.1 | - | 2 - 3 | 90 |