The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024326.3 | NP_077302.3 | 1 - 2 | 1368 | 105 |
XM_005270149.2 | XP_005270206.1 | 3 - 4 | 712 | 105 | |
XM_005270150.4 | XP_005270207.1 | 2 - 3 | 1162 | 105 | |
XM_005270151.3 | XP_005270208.1 | 2 - 3 | 318 | 105 | |
XM_017016631.1 | XP_016872120.1 | 2 - 3 | 449 | 105 | |
GenBank mRNA | AK122684.1 | - | 2 - 3 | 1464 | 105 |
BC002912.2 | - | 1 - 2 | 185 | 105 | |
BC036120.1 | - | 1 - 2 | 1350 | 105 | |
BC130566.1 | - | 2 - 3 | 421 | 105 | |
BI914195.1 | - | 2 - 3 | 615 | 105 |