Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTSSA
Entrez Gene ID
171546
Gene Name
serine palmitoyltransferase small subunit A
Gene Aliases
C14orf147, SSSPTA
Chromosome Location
Chr.14: 34432938 - 34462262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138288.3 NP_612145.2 1 - 2 166 69
GenBank mRNA AK001993.1 - 1 - 2 147 69
AK315409.1 - 1 - 2 144 69
AL080066.1 - 1 - 2 147 69
BC016805.1 - 1 - 2 148 69
BC016808.1 - 1 - 2 148 69
BC021701.1 - 1 - 2 147 69
BC068480.1 - 1 - 2 260 69

View Full Product Details