Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AGAP5
- Entrez Gene ID
-
729092
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 5
- Gene Aliases
-
CTGLF2
- Chromosome Location
-
Chr.10: 73674275 - 73697796 on Build GRCh38
- Gene Symbol
- AGAP12P
- Entrez Gene ID
-
414224
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 12, pseudogene
- Gene Aliases
-
CTGLF12P
- Chromosome Location
-
Chr.10: 48010132 - 48031640 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029396.2 |
|
4 - 5 |
444 |
98 |
- Gene Symbol
- AGAP9
- Entrez Gene ID
-
642517
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 9
- Gene Aliases
-
AGAP-9, CTGLF6, bA301J7.2
- Chromosome Location
-
Chr.10: 47501854 - 47523638 on Build GRCh38
View Full Product Details