Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FKBP1A
- Entrez Gene ID
-
2280
- Gene Name
- FK506 binding protein 1A
- Gene Aliases
-
FKBP-12, FKBP-1A, FKBP1, FKBP12, PKC12, PKCI2, PPIASE
- Chromosome Location
-
Chr.20: 1368977 - 1393172 on Build GRCh38
- Gene Symbol
- FKBP1A-SDCBP2
- Entrez Gene ID
-
100528031
- Gene Name
- FKBP1A-SDCBP2 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 1309909 - 1393172 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037661.1 |
|
1 - 2 |
214 |
63 |
GenBank mRNA |
AK309026.1 |
- |
1 - 2 |
128 |
63 |
BM925547.1 |
- |
1 - 2 |
122 |
63 |
View Full Product Details