Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GTF2IRD2
- Entrez Gene ID
-
84163
- Gene Name
- GTF2I repeat domain containing 2
- Gene Aliases
-
FP630, GTF2IRD2 alpha, GTF2IRD2A
- Chromosome Location
-
Chr.7: 74796144 - 74851576 on Build GRCh38
- Gene Symbol
- GTF2IRD2B
- Entrez Gene ID
-
389524
- Gene Name
- GTF2I repeat domain containing 2B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 75092556 - 75149817 on Build GRCh38
- Gene Symbol
- GTF2IRD2P1
- Entrez Gene ID
-
401375
- Gene Name
- GTF2I repeat domain containing 2 pseudogene 1
- Gene Aliases
-
GTF2IRD2P
- Chromosome Location
-
Chr.7: 73242751 - 73280119 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY312852.1 |
- |
5 - 6 |
429 |
69 |
View Full Product Details