Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SPATA31A7
- Entrez Gene ID
-
26165
- Gene Name
- SPATA31 subfamily A member 7
- Gene Aliases
-
AEP1, C9orf36A, FAM75A4, FAM75A7, SPATA31A4
- Chromosome Location
-
Chr.9: 61190036 - 61196280 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_015667.2 |
NP_056482.2 |
1 - 2 |
220 |
74 |
- Gene Symbol
- SPATA31A1
- Entrez Gene ID
-
647060
- Gene Name
- SPATA31 subfamily A member 1
- Gene Aliases
-
C9orf36, C9orf36A, FAM75A1, FAM75A2, SPATA31A2
- Chromosome Location
-
Chr.9: 39355667 - 39361962 on Build GRCh38
- Gene Symbol
- SPATA31A3
- Entrez Gene ID
-
727830
- Gene Name
- SPATA31 subfamily A member 3
- Gene Aliases
-
FAM75A3
- Chromosome Location
-
Chr.9: 66986301 - 66992550 on Build GRCh38
- Gene Symbol
- SPATA31A5
- Entrez Gene ID
-
727905
- Gene Name
- SPATA31 subfamily A member 5
- Gene Aliases
-
FAM75A5
- Chromosome Location
-
Chr.9: 60914372 - 60920653 on Build GRCh38
- Gene Symbol
- SPATA31A6
- Entrez Gene ID
-
389730
- Gene Name
- SPATA31 subfamily A member 6
- Gene Aliases
-
FAM75A6
- Chromosome Location
-
Chr.9: 42183659 - 42189887 on Build GRCh38
View Full Product Details