Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRAMEF12
Entrez Gene ID
390999
Gene Name
PRAME family member 12
Gene Aliases
-
Chromosome Location
Chr.1: 12774848 - 12777906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080830.4 NP_001074299.2 1 - 2 312 94

View Full Product Details