Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRH2
Entrez Gene ID
5555
Gene Name
proline rich protein HaeIII subfamily 2
Gene Aliases
PRP-1/PRP-2, Pr, pr1/Pr2
Chromosome Location
Chr.12: 10929236 - 10934845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC136499.1 - 3 - 3 644 102
BX641094.1 - 3 - 3 657 102
BX647155.1 - 4 - 4 551 102
BX648656.1 - 3 - 3 583 102
BX648871.1 - 2 - 2 628 102

View Full Product Details