The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006830.3 | NP_006821.1 | 1 - 1 | 54 | 89 |
GenBank mRNA | DW465790.1 | - | 1 - 1 | 59 | 89 |