The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000165.4 | NP_000156.1 | 2 - 2 | 974 | 92 |
GenBank mRNA | AK223289.1 | - | 2 - 2 | 905 | 92 |
AK294670.1 | - | 3 - 3 | 673 | 92 | |
AK295460.1 | - | 3 - 3 | 819 | 92 | |
AK297402.1 | - | 2 - 2 | 666 | 92 | |
AK297764.1 | - | 3 - 3 | 858 | 92 | |
AK312324.1 | - | 2 - 2 | 916 | 92 | |
BC026329.1 | - | 2 - 2 | 790 | 92 | |
CR541660.1 | - | 1 - 1 | 702 | 92 | |
M65188.1 | - | 2 - 2 | 867 | 92 | |
X52947.1 | - | 2 - 2 | 859 | 92 |