The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000862.2 | NP_000853.1 | 4 - 4 | 752 | 112 |
XM_011541314.2 | XP_011539616.1 | 4 - 4 | 793 | 112 | |
XM_017001119.1 | XP_016856608.1 | 4 - 4 | 716 | 112 | |
GenBank mRNA | AK291556.1 | - | 3 - 3 | 766 | 112 |
AL544564.3 | - | 4 - 4 | 752 | 112 | |
AL550307.3 | - | 4 - 4 | 734 | 112 | |
BC031999.1 | - | 3 - 3 | 694 | 112 | |
M27137.1 | - | 3 - 3 | 610 | 112 | |
M35493.1 | - | 3 - 3 | 637 | 112 | |
S45679.1 | - | 4 - 4 | 738 | 112 | |
X53321.1 | - | 3 - 3 | 637 | 112 | |
X55997.1 | - | 3 - 3 | 607 | 112 |