The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004308.2 | NP_001004308.2 | 3 - 3 | 2252 | 107 |
GenBank mRNA | AK128465.1 | - | 3 - 3 | 2251 | 107 |
BC137430.1 | - | 1 - 1 | 2044 | 107 | |
BC137433.1 | - | 1 - 1 | 2044 | 107 |