The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017528.2 | NP_001017528.1 | 9 - 9 | 1314 | 66 |
NM_001017529.2 | NP_001017529.1 | 8 - 8 | 1197 | 66 | |
NM_001017530.1 | NP_001017530.1 | 8 - 8 | 1371 | 66 | |
NM_001198721.1 | NP_001185650.1 | 10 - 10 | 2027 | 66 | |
NM_015366.3 | NP_056181.2 | 9 - 9 | 1488 | 66 | |
NM_181333.3 | NP_851850.1 | 8 - 8 | 1349 | 66 | |
GenBank mRNA | AF177331.1 | - | 8 - 8 | 1293 | 66 |
AK000192.1 | - | 4 - 4 | 1407 | 66 | |
AK055848.1 | - | 10 - 10 | 1750 | 66 | |
AK056787.1 | - | 2 - 2 | 1435 | 66 | |
AK091884.1 | - | 9 - 9 | 1239 | 66 | |
AK291564.1 | - | 8 - 8 | 1339 | 66 | |
BC016921.1 | - | 4 - 4 | 1596 | 66 |