The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098668.3 | NP_001092138.1 | 6 - 6 | 1603 | 93 |
NM_001320813.1 | NP_001307742.1 | 6 - 6 | 1606 | 93 | |
NM_001320814.1 | NP_001307743.1 | 5 - 5 | 1672 | 93 | |
XM_005270128.3 | XP_005270185.1 | 6 - 6 | 1584 | 93 | |
XM_005270132.3 | XP_005270189.1 | 5 - 5 | 1571 | 93 | |
XM_011540124.1 | XP_011538426.1 | 6 - 6 | 1699 | 93 | |
XM_011540125.1 | XP_011538427.1 | 6 - 6 | 1642 | 93 | |
XM_017016608.1 | XP_016872097.1 | 5 - 5 | 1495 | 93 | |
GenBank mRNA | BC139727.1 | - | 4 - 4 | 1475 | 93 |
HQ021421.1 | - | 6 - 6 | 1552 | 93 | |
HQ021422.2 | - | 6 - 6 | 1552 | 93 | |
HQ021423.2 | - | 6 - 6 | 1552 | 93 | |
HQ021424.1 | - | 6 - 6 | 1552 | 93 | |
HQ021425.2 | - | 6 - 6 | 1552 | 93 | |
HQ021426.1 | - | 6 - 6 | 1549 | 93 | |
HQ021427.1 | - | 6 - 6 | 1549 | 93 | |
HQ021428.2 | - | 6 - 6 | 1549 | 93 | |
HQ021429.2 | - | 6 - 6 | 1549 | 93 | |
HQ021430.1 | - | 6 - 6 | 1549 | 93 | |
HQ021431.2 | - | 6 - 6 | 1549 | 93 | |
HQ021432.1 | - | 6 - 6 | 1552 | 93 |