The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142448.1 | NP_001135920.1 | 13 - 13 | 1935 | 74 |
NM_001142449.1 | NP_001135921.1 | 10 - 10 | 1950 | 74 | |
NM_001142450.1 | NP_001135922.1 | 13 - 13 | 2085 | 74 | |
NM_001142451.1 | NP_001135923.1 | 11 - 11 | 2016 | 74 | |
NM_032038.2 | NP_114427.1 | 12 - 12 | 2172 | 74 | |
XM_006721096.3 | XP_006721159.1 | 12 - 12 | 1786 | 74 | |
XM_017023758.1 | XP_016879247.1 | 13 - 13 | 1954 | 74 | |
XM_017023759.1 | XP_016879248.1 | 13 - 13 | 1938 | 74 | |
GenBank mRNA | BC065235.1 | - | 13 - 13 | 1917 | 74 |
CD368853.1 | - | 1 - 1 | 74 |