Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NANOGNB
Entrez Gene ID
360030
Gene Name
NANOG neighbor homeobox
Gene Aliases
-
Chromosome Location
Chr.12: 7765216 - 7774121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145465.1 NP_001138937.1 2 - 2 448 80
GenBank mRNA AY151139.1 - 2 - 2 448 80

View Full Product Details