Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ADAM20P1
- Entrez Gene ID
-
317760
- Gene Name
- ADAM metallopeptidase domain 20 pseudogene 1
- Gene Aliases
-
C14orf55
- Chromosome Location
-
Chr.14: 70468876 - 70483756 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037933.1 |
|
2 - 2 |
1453 |
100 |
GenBank mRNA |
BC047939.1 |
- |
2 - 2 |
1453 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ADAM metallopeptidase domain 21 |
ADAM21 |
ADAM 21, ADAM31 |
-9.0kb
|
Anti-sense |
Chr. - 70452157 - 70459905 on Build GRCh38 |
Downstream |
ADAM metallopeptidase domain 20 |
ADAM20 |
|
49.3kb
|
Sense |
Chr. - 70520817 - 70564742 on Build GRCh38 |
View Full Product Details