The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139208.2 | NP_631947.1 | 4 - 5 | 581 | 69 |
GenBank mRNA | AB008047.1 | - | 4 - 5 | 575 | 69 |