The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100879.1 | NP_001094349.1 | 3 - 3 | 837 | 104 |
NM_001100880.2 | NP_001094350.1 | 3 - 3 | 370 | 104 | |
NM_001199757.1 | NP_001186686.1 | 3 - 3 | 348 | 104 | |
NR_037652.1 | 3 - 3 | 471 | 104 | ||
NR_037653.1 | 3 - 3 | 493 | 104 | ||
GenBank mRNA | AA282293.1 | - | 3 - 3 | 395 | 104 |
AF249277.1 | - | 3 - 3 | 439 | 104 | |
AL109714.1 | - | 3 - 3 | 837 | 104 | |
BM559302.1 | - | 3 - 3 | 460 | 104 | |
BQ680929.1 | - | 3 - 3 | 452 | 104 |