Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGFR1
Entrez Gene ID
2260
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Chromosome Location
Chr.8: 38411138 - 38468834 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011544452.2 XP_011542754.1 7 - 8 1261 100
XM_017013229.1 XP_016868718.1 1 - 2 125 100
XM_017013230.1 XP_016868719.1 5 - 6 731 100
XM_017013231.1 XP_016868720.1 7 - 8 1271 100
GenBank mRNA FJ809916.1 - 7 - 8 1875 100
FJ809917.1 - 7 - 8 1875 100

View Full Product Details