The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198277.2 | NP_938018.1 | 17 - 18 | 1741 | 95 |
GenBank mRNA | AK296975.1 | - | 7 - 8 | 714 | 95 |
BC051314.1 | - | 17 - 18 | 1522 | 95 |