Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CYP4F24P
- Entrez Gene ID
-
388514
- Gene Name
- cytochrome P450 family 4 subfamily F member 24, pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 15759334 - 15779988 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033864.1 |
|
3 - 4 |
460 |
102 |
GenBank mRNA |
AK308750.1 |
- |
3 - 4 |
455 |
102 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CD558184.1 |
- |
|
484 |
|
Chr. - - [-] on Build |
102 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
olfactory receptor family 10 subfamily H member 3 |
OR10H3 |
|
-30.3kb
|
Anti-sense |
Chr. - 15741393 - 15742343 on Build GRCh38 |
Downstream |
olfactory receptor family 10 subfamily H member 5 |
OR10H5 |
OR19-25, OR19-26 |
19.9kb
|
Anti-sense |
Chr. - 15794049 - 15794996 on Build GRCh38 |
View Full Product Details