The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244959.1 | NP_001231888.1 | 14 - 15 | 1859 | 82 |
NM_001244961.1 | NP_001231890.1 | 9 - 10 | 1171 | 82 | |
NM_001244962.1 | NP_001231891.1 | 2 - 3 | 738 | 82 | |
GenBank mRNA | AK094281.1 | - | 14 - 15 | 1560 | 82 |
BC023560.1 | - | 2 - 3 | 738 | 82 | |
BC037253.1 | - | 9 - 10 | 1171 | 82 |